Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTEX1D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534446
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534446 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534446 Supplier Invitrogen™ Supplier No. PA534446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is EL4 cell lysate. PA5-34446 can be used with blocking peptide PEP-1488.

Dyneins are multi-subunit, high molecular weight ATPases that interact with microtubules to generate force by converting the chemical energy of ATP into the mechanical energy of movement. Cytoplasmic or axonemal Dynein is an approximately 12 subunit complex of two heavy chains, two intermediate chains to anchor Dynein to its cargo, four smaller intermediate chains and several light chains. It performs functions necessary for cell survival such as organelle transport and centrosome assembly. TCTEX1 is a cytoplasmic dynein light chain found in a complex with Na+ CP type X (SCN10A). TCTEX1D3, also called TCTE3, has been identified as a gene responsible for congenital diaphragmatic hernia (CDH).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTEX1D3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TCTE3
Gene Accession No. P11985, Q8IZS6
Gene Alias D17Leh117c; LC2; t-complex testis-expressed 4; T-complex testis-specific protein 2; T-complex testis-specific protein 3; t-complex-associated testis expressed 3; T-complex-associated testis-expressed protein 3; T-complex-associated testis-expressed protein 4; t-complex-associated-testis-expressed 3; TCTE3; tcte-3; Tctex1 domain containing 3; tctex1 domain-containing protein 3; Tctex1d3; Tctex2; TCTEX-2; Tctex4; Tctex-4
Gene Symbols TCTE3
Host Species Rabbit
Immunogen A 19 amino acid synthetic peptide near the amino terminus of human TCTEX1D3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21647, 365153, 6991
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.