Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCTEX1D4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534447
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534447 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534447 Supplier Thermo Scientific Supplier No. PA534447
Add to Cart
Add to Cart

TCTEX1D4 Polyclonal Antibody

TCTEX1D4 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTEX1D4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TCTEX1D4
Gene Accession No. Q5JR98, Q8CDY7
Gene Alias 4833401D15Rik; LOC102551208; N22.1; novel Tctex-1 family domain-containing protein; protein N22.1; Tctex1 domain containing 4; tctex1 domain-containing protein 4; tctex1 domain-containing protein 4-like; Tctex1d4; Tctex2 beta; Tctex-2-beta
Gene Symbols TCTEX1D4
Host Species Rabbit
Immunogen A 19 amino acid synthetic peptide near the amino terminus of human TCTEX1D4.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102551208, 242646, 343521
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.