Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590130 Supplier Invitrogen™ Supplier No. PA590130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFTANPPQRV FELVDQINPS IFCIHITNYK PALSFINPEV PDENNFDTLM KTSDGFTLNA ESYVSFTTKL DIPTAAKYEY GVPLQTSDSF LRFPSSLTSS LCTDNNPAAF LVNQAVKCTR KINLEQCEEI EALSMAFYSS PEILRVPDSR KKVPITVQSI VIQSLNKTLT RREDTDVLQP T; Positive Samples: MCF7, HT-29, 293T, NCI-H460; Cellular Location: Cytoplasm, cilium basal body, cytoskeleton.

TCTN1 is the founding member of the Tectonic protein family, a group of evolutionarily conserved secreted and transmembrane proteins that regulate the Hedgehog (Hh)-mediated patterning of the neural tube. During embryonic development, TCTN1 is expressed in regions that participate in Hh signaling, beginning in the gastrulation stages in the ventral node. Mice expressing mutant TCTN1 die between E13. 5 and E16. 5 and display holoprosencephaly, a defect associated with reduced Hh signaling, indicating the role of TCTN1 as an Hh activator. At later stages in development, TCTN1 is thought to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TCTN1
Gene Accession No. Q2MV58
Gene Alias G730031O11Rik; JBTS13; RGD1566266; TCTN1; TECT1; tectonic family member 1; tectonic-1; UNQ9369/PRO34160
Gene Symbols TCTN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 130-310 of human TCTN1 (NP_0010760061).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79600
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.