Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCTN2 Antibody (OTI1C1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274483
Description
TCTN2 Monoclonal specifically detects TCTN2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
TCTN2 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
C12orf38, chromosome 12 open reading frame 38, FLJ12975, TECT2tectonic 2, tectonic family member 2, tectonic-2 | |
Mouse | |
76.7 kDa | |
100 μg | |
79867 | |
Human | |
Lyophilized |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
OTI1C1 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
TCTN2 | |
Full length human recombinant protein of human TCTN2 (NP_079085) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction