Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCTN2 Antibody (OTI1C1), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274483F
Description
TCTN2 Monoclonal specifically detects TCTN2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
TCTN2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TCTN2 | |
Full length recombinant protein of human TCTN2 (NP_079085) produced in HEK293T cells. | |
0.1 mL | |
79867.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1C1 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
C12orf38, chromosome 12 open reading frame 38, FLJ12975, TECT2tectonic 2, tectonic family member 2, tectonic-2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction