Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596985 Supplier Invitrogen™ Supplier No. PA596985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKFLSYNSGN EEELSGNPGY QLGKPVRALN INRMNNVTTL HLWQSAGRGL CTSATFKPIL FGENVLSGCL LEVGINENCT QLRENAVERL DSLIQATHVA MRGNSDYADL SDGWLEIIRV DAPDPGADPL ASSVNGMCLD IPAHLSIRIL ISDAGAVEGI TQQEILGVET RFSSVNWQYQ CGLTCEHKAD LLPISASVQF IKIPAQLPHP LTRFQINYTE YDCNRNEVCW PQLLYPWTQY YQGELHSQCV A; Positive Samples: Mouse testis, Mouse brain, Mouse kidney, Mouse lung; Cellular Location: Cytoplasm, Membrane, Single-pass type I membrane protein, cilium basal body, cytoskeleton.

TCTN2 is a member of the Tectonic protein family, a group of evolutionarily conserved secreted and transmembrane proteins that regulate the Hedgehog (Hh)-mediated patterning of the neural tube. While the expression pattern and function of TCTN2 is not known, it is 49% similar to TCTN1. TCTN1 is expressed during embryonic development in regions that participate in Hh signaling, beginning in the gastrulation stages in the ventral node. Mice expressing mutant TCTN1 die between E13. 5 and E16. 5 and display holoprosencephaly, a defect associated with reduced Hh signaling, indicating the role of TCTN1 as an Hh activator. At later stages in development, TCTN1 is thought to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TCTN2
Gene Accession No. Q2MV57
Gene Alias 4432405B04Rik; C12orf38; JBTS24; MKS8; Tctn2; TECT2; tectonic 2; tectonic family member 2; tectonic-2
Gene Symbols TCTN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 420-670 of human TCTN2 (NP_0790852).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67978
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.