Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDG Antibody, Novus Biologicals™
SDP

Catalog No. NBP232248 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232248 Supplier Novus Biologicals Supplier No. NBP232248
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDG Polyclonal specifically detects TDG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TDG
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2 - 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_003202.3
Gene Alias EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase
Gene Symbols TDG
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 360 to 410 of human Thymine-DNA Glycosylase using the numbering given in entry NP_003202.3 (GeneID 6996).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Base Excision Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 6996
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.