Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDG Antibody (OTI3C11), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274484AF647
Description
TDG Monoclonal specifically detects TDG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TDG | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TDG | |
Recombinant protein fragment corresponding to amino acids 160-253 of human TDG (NP_003202) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3C11 | |
Western Blot, Immunohistochemistry | |
EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase | |
Mouse | |
Affinity Purified | |
Base Excision Repair, DNA Repair, Epigenetics, microRNA, Stem Cell Markers | |
6996.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction