Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDG Monoclonal Antibody (GT1142)
GREENER_CHOICE

Catalog No. PIMA527785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527785 Supplier Invitrogen™ Supplier No. MA527785
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: human TDG -transfected 293T Predicted Reactivity: Mouse (100%), Rat (99%), Xenopus laevis (92%), Chicken (95%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene belongs to the TDG/mug DNA glycosylase family. Thymine-DNA glycosylase removes thymine moieties from G/T mismatches by hydrolyzing the carbon-nitrogen bond between the sugar-phosphate backbone of DNA and the mis paired thymine. With lower activity, this enzyme also removes thymine from C/T and T/T mispairings. TDG can also remove uracil and 5-bromouracil from mispairings with guanine. This enzyme plays a central role in cellular defense against genetic mutation caused by the spontaneous deamination of 5-methylcytosine and cytosine. This gene may have a pseudogene in the p arm of chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDG
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT1142
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TDG
Gene Accession No. Q13569
Gene Alias C-JUN leucine zipper interactive protein JZA-3; E130317C12Rik; G/T mismatch-specific thymine DNA glycosylase; hTDG; Jza1; JZA-3; Tdg; thymine DNA glycosylase; thymine-DNA glycosylase
Gene Symbols TDG
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human TDG. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.