Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596415
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596415 Supplier Invitrogen™ Supplier No. PA596415
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-29, MCF7, HL-60, Mouse stomach, rat eye, rat stomach; Cellular Location: Nucleus Immunogen sequence: MEAENAGSYS LQQAQAFYTF PFQQLMAEAP NMAVVNEQQM PEEVPAPAPA QEPVQEAPKG RKRKPRTTEP KQPVEPKKPV ESKKSGKSAK SKEKQEKITD TFKVKRKVDR FNGVSEAELL TKTLPDILTF NLDIVIIGIN PGLMAAYKGH HYPGPGNHFW.

The protein encoded by this gene belongs to the TDG/mug DNA glycosylase family. Thymine-DNA glycosylase removes thymine moieties from G/T mismatches by hydrolyzing the carbon-nitrogen bond between the sugar-phosphate backbone of DNA and the mis paired thymine. With lower activity, this enzyme also removes thymine from C/T and T/T mispairings. TDG can also remove uracil and 5-bromouracil from mispairings with guanine. This enzyme plays a central role in cellular defense against genetic mutation caused by the spontaneous deamination of 5-methylcytosine and cytosine. This gene may have a pseudogene in the p arm of chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TDG
Gene Accession No. P56581, Q13569
Gene Alias C-JUN leucine zipper interactive protein JZA-3; E130317C12Rik; G/T mismatch-specific thymine DNA glycosylase; hTDG; Jza1; JZA-3; Tdg; thymine DNA glycosylase; thymine-DNA glycosylase
Gene Symbols TDG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human TDG (NP_0032023).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114521, 21665, 6996
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.