Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDI Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TDI |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TDI Polyclonal specifically detects TDI in Human samples. It is validated for Western Blot.Specifications
TDI | |
Polyclonal | |
Rabbit | |
Human | |
100038246 | |
Synthetic peptides corresponding to TD1(TLX1 divergent) The peptide sequence was selected from the middle region of TD1. Peptide sequence PPHPLNKQKHHPPHPSQTQKDLVPRSPQLEKSRIRLRRTLRNLGGGRGQR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
APT-B7, Putative protein TDI, putative TLX1 neighbor protein, TD1, TDI, TLX1 divergent gene protein, TLX1 neighbor | |
TLX1NB | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title