Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDIF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560731 Supplier Invitrogen™ Supplier No. PA560731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSQESHTEAI SDAETSSSDI SFSGIATRRT RSMQRKLKAQ TEKKDSKIVP GNEKQIVGTP VNSEDSDTRQ TSHLQARSLS EINKPNFYNN DFDDDFSHR Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 55%.

This gene is thought to be involved in chromatin remodeling and gene transcription. The encoded nuclear protein binds to and enhances the transcriptional activity of the estrogen receptor alpha, and also interacts with terminal deoxynucleotidyltransferase. The expression profile of this gene is a potential biomarker for chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDIF2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNTTIP2
Gene Accession No. Q5QJE6
Gene Alias 4930588M11Rik; AA408582; acidic 82 kDa protein mRNA; AU014960; deoxynucleotidyltransferase terminal interacting protein 2; deoxynucleotidyltransferase terminal-interacting protein 2; deoxynucleotidyltransferase, terminal, interacting protein 2; DNTTIP2; ERBP; estrogen receptor binding protein; Estrogen receptor-binding protein; FCF2; HSU15552; LPTS-interacting protein 2; LPTS-RP2; TDIF2; tdT-interacting factor 2; Terminal deoxynucleotidyltransferase-interacting factor 2
Gene Symbols DNTTIP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TDIF2. Recombinant protein control fragment (Product #RP-94005).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.