Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDIF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571407 Supplier Invitrogen™ Supplier No. PA571407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDIF2 Polyclonal Antibody for Western Blot

This gene is thought to be involved in chromatin remodeling and gene transcription. The encoded nuclear protein binds to and enhances the transcriptional activity of the estrogen receptor alpha, and also interacts with terminal deoxynucleotidyltransferase. The expression profile of this gene is a potential biomarker for chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDIF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNTTIP2
Gene Accession No. Q5QJE6
Gene Alias 4930588M11Rik; AA408582; acidic 82 kDa protein mRNA; AU014960; deoxynucleotidyltransferase terminal interacting protein 2; deoxynucleotidyltransferase terminal-interacting protein 2; deoxynucleotidyltransferase, terminal, interacting protein 2; DNTTIP2; ERBP; estrogen receptor binding protein; Estrogen receptor-binding protein; FCF2; HSU15552; LPTS-interacting protein 2; LPTS-RP2; TDIF2; tdT-interacting factor 2; Terminal deoxynucleotidyltransferase-interacting factor 2
Gene Symbols DNTTIP2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human TDIF2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30836
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.