Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDO2, Mouse anti-Human, Clone: 4G2, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16405
Specifications
TDO2 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide). | |
TDO/TPH2/TRPO | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
4G2 | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
tryptophan 2,3-dioxygenase | |
TDO2 | |
Recombinant protein corresponding to full length human TDO2 from HEK293T cell. | |
RUO | |
6999 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction