Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TDO2, Mouse, Unlabeled, Clone: 7A6.1,

Catalog No. MABN1537MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1537MI 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1537MI Supplier MilliporeSigma™ Supplier No. MABN1537
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E3 ubiquitin-protein ligase RNF43 (UniProt Q68DV7; also known as RING finger protein 43) is encoded by the RNF43 gene (Gene ID 54894) in human. RNF43 and the homologous ZNRF3 are cell-surface transmembrane E3 ubiquitin ligases upregulated upon Wnt/ -catenin signaling and act as potent Wnt signaling pathway negative feedback regulators by mediating ubiquitination of Frizzled (FZD) and LRP6 for lysosomal degradation. RNF43/ZNRF3 activity is counteracted by stem cell growth factor R-spondin that induces membrane ZNRF3/RNF43 clearance by binding ZNRF3/RNF43 and LGR4/LGR5. Physical interaction with Dishevelled (DVL) is essential for ZNRF3/RNF43 Wnt inhibitory activity, and DVL knockdown significantly increases cell surface levels of FZD and LRP6. In addition, RNF43 suppresses canonical and noncanonical Wnt signaling via distinct mechanisms. While interaction between the extracellular protease-associated (PA) domain and the cysteine-rich domain (CRD) of FZD and RNF43 intracellular RING finger domain is necessary for canonical Wnt/ -catenin signaling inhibition, interaction between FZD PDZ domain and RNF43 C-terminal cytoplasmic region is essential for suppressing noncanonical Wnt signaling. R-spondin overexpression through gene translocation and loss-of-function RNF43/ZNRF3 gene mutations are found in human cancers as means to escape from Wnt signaling negative feedback regulation. Human RNF43 is produced with a signal peptide (a.a. 1-23) sequence, which is posttranslationally removed to yiled the mature enzyme with an N-terminal extracellular domain (a.a. 24-197), a transmembrane domain (a.a. 198-218), and a large cytoplasmic region (a.a. 219-783) that contains an atypical RING-type zinc finger (a.a. 272-313) and a pro-rich (a.a. 569-760) region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDO2
Applications Western Blot
Classification Monoclonal
Clone 7A6.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene TDO2, TDO
Gene Accession No. P48775
Host Species Mouse
Immunogen GST-tagged recombinant human TDO2 internal fragment.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005642
Test Specificity Clone 7A6.1 targets an internal epitope of human and murine TDO2 (TDO).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.