Learn More
Invitrogen™ TDP1 Monoclonal Antibody (2A10-G2)

Description
Peptide Sequence: MSQEGDYGRW TISSSDESEE EKPKPDKPST SSLLCARQGA ANEPRYTCSE AQKAAHKRKI SPVKFSNTDS VLPPKRQKSG SQEDLGWCLS SSDDELQPEM PQKQAEKVVI KKEKDISAPN DGTAQRTENH GAPACHRLKE EEDEYETSGE GQDIWDMLDK GNPFQFYLTR VSGVKPKYNS GALHIKDILS PLFGTLVSSA QFNYCFDVDW LVKQYPPEFR KKPILLVHGD KREAKAHLHA QAKPYENISL CQAKLDIAFG THHTKMMLLL YEEGLRVVIH TSNLIHADWH QKTQGIWLSP LYPRIADGTH KSGESPTHFK ADLISYLMAY NAPSLKEWID VIHKHDLSET NVYLIGSTPG RFQGSQKDNW GHFRLKKLLK DHASSMPNAE SWPVVGQFSS VGSLGADESK WLCSEFKESM LTLGKESKTP GKSSVPLYLI YPSVENVRTS LEGYPAGGSL PYSIQTAEKQ NWLHSYFHKW SAETSGRSNA MPHIKTYMRP SPDFSKIAWF LVTSANLSKA AWGALEKNGT QLMIRSYELG VLFLPSAFGL DSFKVKQKFF AGSQEPMATF PVPYDLPPEL YGSKDRPWIW NIPYVKAPDT HGNMWVPS.
Specifications
Specifications
| Antigen | TDP1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2A10-G2 |
| Concentration | 0.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Tdp1 |
| Gene Accession No. | Q9NUW8 |
| Gene Alias | 2810481F14Rik; 4921509N21Rik; AI838772; AW493413; E430034L06Rik; MLZ-501; protein expressed in male leptotene and zygotene spermatocytes 501; SCAN1; Tdp1; Tyr-DNA phosphodiesterase 1; tyrosyl-DNA phodphodiesterase 1; tyrosyl-DNA phosphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1-like protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.