Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD1 Monoclonal Antibody (739206)
GREENER_CHOICE

Catalog No. PIMA524351
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524351 Supplier Invitrogen™ Supplier No. MA524351
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

This gene is similar to a mouse gene that encodes a tudor domain protein. Alternatively spliced transcript variants have been described but their full length sequences have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD1
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 739206
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Tdrd1
Gene Accession No. Q99MV1
Gene Alias Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1
Gene Symbols Tdrd1
Host Species Rat
Immunogen E. coli-derived recombinant mouse TDRD1 Asn831-Lys960.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83561
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.