Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558079 Supplier Invitrogen™ Supplier No. PA558079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TENNIVVDKS DLIPKVLTLN VGDEFCGVVA HIQTPEDFFC QQLQSGRKLA ELQASLSKYC DQLPPRSDFY PAIGDICCAQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.

This gene is similar to a mouse gene that encodes a tudor domain protein. Alternatively spliced transcript variants have been described but their full length sequences have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tdrd1
Gene Accession No. Q9BXT4
Gene Alias Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1
Gene Symbols Tdrd1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TDRD1. Recombinant protein control fragment (Product #RP-97027).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56165
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.