Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588834 Supplier Invitrogen™ Supplier No. PA588834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFNFEKNENK LPPHESLRSP GTLPNHPNFR LKSSENGNKK NNFLLCEQTK QYLASQEDNS VSSNPNGING EVVGSKGDRK KLPAGNSVSP PSAESNSPPK E; Positive Samples: Mouse testis; Cellular Location: Cytoplasm.

This gene is similar to a mouse gene that encodes a tudor domain protein. Alternatively spliced transcript variants have been described but their full length sequences have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Tdrd1
Gene Accession No. Q99MV1
Gene Alias Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1
Gene Symbols Tdrd1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-125 of human TDRD1 (NP_9420901).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83561
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.