Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209338
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9338 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9338 Supplier Fabgennix Inc Supplier No. TDRD1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRD1 Polyclonal antibody specifically detects TDRD1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen TDRD1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Tdrd1
Gene Accession No. Q99MV1, Q9BXT4
Gene Alias Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 570-605 on TDRD1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292129, 56165, 83561
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.