Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144251
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144251 Supplier Invitrogen™ Supplier No. PA5144251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRD12 Polyclonal Antibody for ELISA, IHC (P)

TDRD12 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD12
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TDRD12
Gene Accession No. Q587J7
Gene Alias ECAT8; ES cell associated transcript 8; ES cell-associated transcript 8 protein; Putative ATP-dependent RNA helicase TDRD12; TDRD12; tudor domain containing 12; tudor domain-containing protein 12
Gene Symbols TDRD12
Host Species Rabbit
Immunogen Recombinant Human Putative ATP-dependent RNA helicase TDRD12 protein (991-1164AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91646
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.