Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146820AF405
Description
TDRD3 Polyclonal antibody specifically detects TDRD3 in Human samples. It is validated for Western BlotSpecifications
TDRD3 | |
Polyclonal | |
Western Blot | |
FLJ21007, tudor domain containing 3, tudor domain-containing protein 3 | |
The immunogen for this product maps to a region between residue 400 and 450 of human tudor domain containing 3 using the numbering given in entry NP_110421.1 (GeneID 81550). | |
0.1 mL | |
Neurodegeneration, Neuroscience | |
81550 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction