Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD3 Antibody (RAB-C419), DyLight 755 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320149IR
Description
TDRD3 Monoclonal antibody specifically detects TDRD3 in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
TDRD3 | |
Monoclonal | |
DyLight 755 | |
FLJ21007, tudor domain containing 3, tudor domain-containing protein 3 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing TDRD3 protein under non-denaturing conditions. | |
0.1 mL | |
Neurodegeneration, Neuroscience | |
81550 | |
Store at 4°C in the dark. | |
IgG κ |
ELISA, Immunofluorescence | |
RAB-C419 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction