Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584749 Supplier Invitrogen™ Supplier No. PA584749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRFQRDSQNS KSVLEGSGLP RNRGSERPST SSVSEVWAED RIKCDRPYSR YDRTKDTSYP LGSQHSDGAF KKRDNSMQSR SGKGP Highest antigen sequence indentity to the following orthologs: Mouse - 75%, Rat - 80%.

TDRD3 belongs to the tudor domain group of proteins, which are prevalent among proteins involved in RNA metabolism. TDRD3 is widely expressed and localizes predominantly in the cytoplasm. It is detected in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Overexpression of TDRD3 in cells induces the formation of stress granules. Multiple isoforms of TDRD3 exist due to alternative splicing variants. In humans, the gene is located on chromosome 13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD3
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TDRD3
Gene Accession No. Q9H7E2
Gene Alias 4732418C03Rik; 6720468N18; Tdrd3; tudor domain containing 3; tudor domain-containing protein 3
Gene Symbols TDRD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TDRD3. Recombinant protein control fragment (Product #RP-108522).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.