Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114394
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114394 Supplier Invitrogen™ Supplier No. PA5114394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human HL-60 whole cell, human Hela whole cell, human PC-3 whole cell, human HEK293 whole cell, human A549 whole cell, human A431 whole cell, rat brain tissues, rat liver tissue, mouse brain tissue, mouse lung tissue, mouse liver tissue, mouse NIH3T3 whole cell. IHC: human colon cancer tissue, mouse small intestine tissue, rat small intestine tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

TDRD3 belongs to the tudor domain group of proteins, which are prevalent among proteins involved in RNA metabolism. TDRD3 is widely expressed and localizes predominantly in the cytoplasm. It is detected in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Overexpression of TDRD3 in cells induces the formation of stress granules. Multiple isoforms of TDRD3 exist due to alternative splicing variants. In humans, the gene is located on chromosome 13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene TDRD3
Gene Accession No. Q66HC1, Q91W18, Q9H7E2
Gene Alias 4732418C03Rik; 6720468N18; Tdrd3; tudor domain containing 3; tudor domain-containing protein 3
Gene Symbols TDRD3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human TDRD3 (KPTNDNDEFEKQRTAAIAEVAKSKETK).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219249, 306066, 81550
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.