Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572627
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2627 100 μL
50-157-2628 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2627 Supplier Bethyl Laboratories, Inc Supplier No. IHC00737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Rabbit, Pig, Panda, Orangutan

TDRD3 belongs to the tudor domain group of proteins, which are prevalent among proteins involved in RNA metabolism. TDRD3 is widely expressed and localizes predomitly in the cytoplasm. It is detected in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Overexpression of TDRD3 in cells induces the formation of stress granules. Multiple isoforms of TDRD3 exist due to alternative splicing varients. In humans, the gene is located on chromosome 13.

Specifications

Antigen TDRD3
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TDRD3
Gene Accession No. Q9H7E2
Gene Alias Tudor domain-containing protein 3
Gene Symbols TDRD3
Host Species Rabbit
Immunogen Between 601 and 651
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81550
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.