Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563229
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3229 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3229 Supplier Bethyl Laboratories, Inc Supplier No. A302691AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TDRD3 belongs to the tudor domain group of proteins, which are prevalent among proteins involved in RNA metabolism. TDRD3 is widely expressed and localizes predomitly in the cytoplasm. It is detected in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Overexpression of TDRD3 in cells induces the formation of stress granules. Multiple isoforms of TDRD3 exist due to alternative splicing varients. In humans, the gene is located on chromosome 13.

Specifications

Antigen TDRD3
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TDRD3
Gene Accession No. Q9H7E2
Gene Alias Tudor domain-containing protein 3
Gene Symbols TDRD3
Host Species Rabbit
Immunogen Between 400 and 450
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81550
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.