Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728677 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8677 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8677 Supplier Proteintech Group Inc Supplier No. 133591AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tudor domain-containing protein 3 (TDRD3) bears one tudor domain and an ubiquitin associated domain (UBA). Tudor domains interact with arginine-glycine-rich motifs in a methylarginine-dependent manner. TDRD3 has been shown to localize to cytoplasmic stress granules and may play a role in the regulation of mRNA translation.

Specifications

Antigen TDRD3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TDRD3
Gene Accession No. Q66HC1, Q91W18, Q9H7E2
Gene Alias TDRD3, tudor domain containing 3
Gene Symbols TDRD3
Host Species Rabbit
Immunogen TDRD3 Fusion Protein Ag4174
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219249, 306066, 81550
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.