Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556139 Supplier Invitrogen™ Supplier No. PA556139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AMANHDIPPD AVPNKKLCRL PPLDTSSLIG VFVEYIISPS QFYIRIYSRD SSELLEDMMI EMRRCYSNQL VSDRYVMPEC FIQPGHLCCV RISEDKWWYR VIIHRVLEKQ EVEVFYPDFG NIGIVQKSSL RFLKCCYTK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

TDRD5, also named as Tudor domain-containing protein 5, is a 981 amino acid protein, which contains one Tudor domain and three HTH OST-type domains. TDRD5 localizes in the cytoplasm and belongs to the TDRD5 family. TDRD5 is required during spermiogenesis to participate in the repression transposable elements and prevent their mobilization, which is essential for the germline integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TDRD5
Gene Accession No. Q8NAT2
Gene Alias Gm103; Tdrd5; testicular tissue protein Li 194; tudor domain containing 5; tudor domain containing protein 5; tudor domain-containing protein 5; TUDOR3
Gene Symbols TDRD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TDRD5. Recombinant protein control fragment (Product #RP-90812).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.