Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140349
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140349 Supplier Invitrogen™ Supplier No. PA5140349
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VSVVHTNKIG RLDLVNAILP GLCIHCSLQG FEVPDNKNSK KMMHYFSQRT SEAAIRCEFV KFQDRWEVIL ADEHGIIADD MISRY Highest antigen sequence indentity to the following orthologs: Mouse - 68%, Rat - 67%.

Tudor domain-containing protein involved in germ cell development, more specifically the formation of chromatoid body (during spermiogenesis), Balbiani body (during oogenesis), germ plasm (upon fertilization), and for proper miRNA expression and spliceosome maturation. Essential for RNA-dependent helicase UPF1 localization to chromatoid body, for UPF1-UPF2 and UPF1-DDX4 interactions which are required for mRNA degradation, using the extended 3' UTR-triggered nonsense-mediated mRNA decay (NMD) pathway. Involved in spliceosome maturation and mRNA splicing in prophase I spermatocytes through interaction with arginine N-methyltransferase PRMT5 and symmetrically arginine dimethylated SNRPB (small nuclear ribonucleoprotein-associated protein).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TDRD6
Gene Accession No. O60522
Gene Alias SPATA36; Tudor Domain Containing 6
Gene Symbols TDRD6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TDRD6. Recombinant protein control fragment (Product #RP-94961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.