Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555102 Supplier Invitrogen™ Supplier No. PA555102
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNCSDCSIKV TKVDETRGIA HVYLFTPKNF PDPHRSINRQ ITNADLWKHQ KDVFLSAISS GADSPNSKNG NMPMSGNTGE NFRKNLTDVI KKSMVDHTSA FSTEELPPPV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.

TDRD7, also known as Tudor domain containing protein 7, is a component of chromatoid bodies in spermatids (named processing bodies or P-bodies in somatic cells). It is found in a mRNP complex together with TDRD1, TDRD6 and DDX4. It is also found in a complex with CABLES1, PCTK2 and PIWIL1. TDRD7 has been shown to interact with histone H3 tri-methylated at K9 in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TDRD7
Gene Accession No. Q8NHU6
Gene Alias 5730495N10Rik; AI447470; CATC4; PCTAIRE2-binding protein; Pctaire2bp; RP11-508D10.1; TDRD7; TRAP; tudor domain containing 7; tudor domain-containing protein 7; tudor repeat associator with PCTAIRE 2; tudor repeat associator with PCTAIRE-2
Gene Symbols TDRD7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TDRD7. Recombinant protein control fragment (Product #RP-93094).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23424
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.