Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TDRD7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570066
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
TDRD7, also known as Tudor domain containing protein 7, is a component of chromatoid bodies in spermatids (named processing bodies or P-bodies in somatic cells). It is found in a mRNP complex together with TDRD1, TDRD6 and DDX4. It is also found in a complex with CABLES1, PCTK2 and PIWIL1. TDRD7 has been shown to interact with histone H3 tri-methylated at K9 in vitro.
Specifications
TDRD7 | |
Polyclonal | |
Unconjugated | |
TDRD7 | |
5730495N10Rik; AI447470; CATC4; PCTAIRE2-binding protein; Pctaire2bp; RP11-508D10.1; TDRD7; TRAP; tudor domain containing 7; tudor domain-containing protein 7; tudor repeat associator with PCTAIRE 2; tudor repeat associator with PCTAIRE-2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
85425 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9R1R4 | |
TDRD7 | |
synthetic peptide directed towards the middle region of human TDRD7 (aa 813-862). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction