Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD9/HIG1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4566 Supplier Bioss Supplier No. BS2185R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRD9 is a protein coding gene.

Specifications

Antigen TDRD9/HIG1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TDRD9
Gene Accession No. Q8NDG6
Gene Alias 4930441E05Rik; ATP-dependent RNA helicase TDRD9; C14orf75; HIG-1; hypoxia-inducible HIG-1; NET54; putative ATP-dependent RNA helicase TDRD9; Tdrd9; tudor domain containing 9; Tudor domain-containing protein 9
Gene Symbols TDRD9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TDRD9/HIG1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122402
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.