Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144169
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144169 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144169 Supplier Invitrogen™ Supplier No. PA5144169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TDRD9 Polyclonal Antibody for IHC (P), ELISA

TDRD9 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD9
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TDRD9
Gene Accession No. Q8NDG6
Gene Alias 4930441E05Rik; ATP-dependent RNA helicase TDRD9; C14orf75; HIG-1; hypoxia-inducible HIG-1; NET54; putative ATP-dependent RNA helicase TDRD9; TDRD9; tudor domain containing 9; tudor domain-containing protein 9
Gene Symbols TDRD9
Host Species Rabbit
Immunogen Recombinant Human Putative ATP-dependent RNA helicase TDRD9 protein (286-585AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122402
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.