Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TDRD9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568740
Description
This target displays homology in the following species: Cow: 77%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 92%.
TDRD9 is a protein coding gene.
Specifications
TDRD9 | |
Polyclonal | |
Unconjugated | |
TDRD9 | |
4930441E05Rik; ATP-dependent RNA helicase TDRD9; C14orf75; HIG-1; hypoxia-inducible HIG-1; NET54; putative ATP-dependent RNA helicase TDRD9; TDRD9; tudor domain containing 9; tudor domain-containing protein 9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
122402 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NDG6 | |
TDRD9 | |
synthetic peptide directed towards the middle region of human TDRD9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction