Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRG1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241197MFV610
Description
TDRG1 Polyclonal antibody specifically detects TDRG1 in Human samples. It is validated for Western Blot, ELISASpecifications
| TDRG1 | |
| Polyclonal | |
| Western Blot, ELISA | |
| LINC00532, Long Intergenic Non-Protein Coding RNA 532, Testis Development Related 1 (Non-Protein Coding) | |
| Antibody was raised against a 17 amino acid peptide near the center of human TDRG1. The immunogen is located within amino acids 40 - 90 of TDRG1. Amino Acid Squence: CIKISSTPPSQSRLDPQI | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 732253 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction