Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534479
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534479 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534479 Supplier Invitrogen™ Supplier No. PA534479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human testis tissue lysate. PA5-34479 can be used with blocking peptide PEP-1521.

The Testis development related protein 1 (TDRG1) is thought to be a testis-specific, developmentally regulated gene. TDRG1 is expressed in mainly in spermatogenic cells in seminiferous tubules of the adult testis, but is not detected in fetal testis. While little is known of the function of TDRG1, it has been postulated to play an important role in human spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRG1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TDRG1
Gene Accession No. Q3Y452
Gene Alias LINC00532; TDRG1; testis development related 1 (non-protein coding)
Gene Symbols TDRG1
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human TDRG1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 732253
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.