Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDRKH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100348 Supplier Invitrogen™ Supplier No. PA5100348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TDRKH.

TDRKH is a protein of unknown function containing one tudor domain, and two KH domains. It has been found to interact with PIWIL1 and PIWIL4 by binding to methylated arginines residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRKH
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TDRKH
Gene Accession No. Q80VL1, Q9Y2W6
Gene Alias 2700091C21Rik; putative RNA binding protein; RGD1311196; RP11-98D18.8; Tdrd2; TDRD7; Tdrkh; tudor and KH domain containing; tudor and KH domain containing protein; tudor and KH domain-containing protein; tudor domain containing 2; tudor domain-containing protein 2
Gene Symbols TDRKH
Host Species Rabbit
Immunogen A synthesized peptide derived from human TDRKH(Accession Q9Y2W6), corresponding to amino acid residues A248-P298.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11022, 310652, 72634
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.