Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TdT Antibody (4B10C3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | TdT |
---|---|
Clone | 4B10C3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61898
![]() |
Novus Biologicals
NBP261898 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
TdT Monoclonal specifically detects TdT in Human samples. It is validated for Western Blot, ELISA.Specifications
TdT | |
1 mg/mL | |
Monoclonal | |
Purified | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
1791 | |
Purified recombinant fragment of human DNTT (AA: 52-192) expressed in E. Coli. | |
Primary | |
58.5 kDa |
4B10C3 | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
Unconjugated | |
Mouse | |
deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
DNTT | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title