Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TdT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | TdT |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP231368SS
![]() |
Novus Biologicals
NBP231368SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP231368
![]() |
Novus Biologicals
NBP231368 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
TdT Polyclonal specifically detects TdT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TdT | |
Western Blot 1.0 ug/ml, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
P04053 | |
1791 | |
Partial recombinant protein made to an internal sequence of human TdT (between residues 50-300) [UniProt# P04053] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
58.5 kDa |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cell Cycle and Replication, Immunology | |
PBS with 0.05% Sodium Azide | |
deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
DNTT | |
IgG | |
Protein A purified | |
Human. Immunogen's sequence similarity with other species: Rat (77%) Mouse (74%), Bovine (78%). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title