Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TdT Monoclonal Antibody (7UNA8U6), eBioscience™, Invitrogen™

Catalog No. 502550878
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-0878 25 μg
14-973-982 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-255-0878 Supplier Invitrogen™ Supplier No. 14973980
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MAb 7UNA8U6 reacts with human, mouse, and rat TdT. TdT, or terminal-deoxynucleotidyl transferase, is a 60 kDa polymerase responsible for the template-independent addition of N-nucleotides at gene segment junctions of developing lymphocytes. Expression is found in the nucleus of immature lymphocytes but not in mature lymphocytes or non-lymphoid cells. TdT plays a critical role during TCR and Ig gene rearrangement and functions to increase the diversity of the lymphocyte repertoire through the addition of N-nucleotides onto the ends of newly formed P-nucleotides. After positive selection, expression of TdT becomes decreased or absent. Expression of TdT has been shown to be present in lymphoma or lymphoblastic leukemia cells.

Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TdT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7UNA8U6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene DNTT
Gene Accession No. P04053, P09838
Gene Alias Deoxynucleotidyltransferase terminal; deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; Dntt; Nucleosidetriphosphate DNA Deoxynucleotidylexotransferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; TDT; Terminal addition enzyme; terminal deoxynucleotidyl transferase; terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; terminal transferase
Gene Symbols DNTT
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1791, 21673, 294051
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.