Learn More
TdT Monoclonal Antibody (TdT-6), FITC, Invitrogen™

Mouse Monoclonal Antibody
$518.00
Specifications
| Antigen | TdT |
|---|---|
| Clone | TdT-6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-113-8133
![]()
|
Invitrogen™
MHTDT012 |
200 μL |
Each of 1 for $518.00
|
|
|||||
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.Specifications
| TdT | |
| Flow Cytometry | |
| FITC | |
| Mouse | |
| PBS with 1% BSA and 0.1% sodium azide | |
| Deoxynucleotidyltransferase terminal; deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; Dntt; Nucleosidetriphosphate DNA Deoxynucleotidylexotransferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; TDT; Terminal addition enzyme; terminal deoxynucleotidyl transferase; terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; terminal transferase | |
| DNTT | |
| IgG1 | |
| purified | |
| Antibody |
| TdT-6 | |
| Monoclonal | |
| Liquid | |
| Human | |
| P04053 | |
| 1791 | |
| Human Terminal Deoxynucleotidyl Transferase (TdT). | |
| Primary | |
| 4°C, store in dark | |
| DNTT |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.