Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tdTomato Mouse anti-Tag, Clone: OTI4B3, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA180148
Description
Tdtomato fluorescent protein.Specifications
| tdTomato | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Tag |
| Western Blot | |
| OTI4B3 | |
| Unconjugated | |
| tdTomato | |
| A tdTomato tagged recombit protein expressed in E.coli. | |
| 100 μL | |
| Primary | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction