Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tdTomato Mouse anti-Tag, Clone: OTI4B3, TrueMAB™, Origene
SDP

Catalog No. 502241583
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-224-1583 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-224-1583 Supplier OriGene Supplier No. CF180148
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

tdTomato Monoclonal antibody specifically detects tdTomato in Tag samples. It is validated for Western Blot

Specifications

Antigen tdTomato
Applications Western Blot
Classification Monoclonal
Clone OTI4B3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative
Gene Alias tdTomato
Host Species Mouse
Immunogen A tdTomato tagged recombinant protein expressed in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.