Learn More
Cytiva TE 77 PWR Semi-Dry Transfer Unit
Catalog No. 45001783 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Amersham ECL Semi Dry Blotter
Catalog No. For Use With (Equipment)
45-001-783 Amersham ECL Semi Dry Blotter
1 options
Catalog No. 45-001-783 Supplier Cytiva Supplier No. 11001342
The TE 77 PWR offers the convenience of a built-in power supply, and a unique automatic stopping feature. The transfer is stopped when buffer gets depleted, preventing overheating.
  • Tested for use with Amersham ECL Western blotting reagents
  • Built-in power supply
  • Use minimal buffer - only enough to saturate blotting papers and membranes
  • Long-lasting perforated platinum-titanium and stainless steel electrodes give uniform, contamination-free transfers
  • Use low current and voltage for transfers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Amersham ECL Semi Dry Blotter
Type TE 77 PWR Semi Dry Transfer Unit