Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TE Buffer, pH 8.0, RNAse Free, PROMO, Thermo Scientific Chemicals

Catalog No. J62745AKPM
Click to view available options
Quantity:
250 mL

Applications

Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8.0 may also be used as a washing buffer. TE buffer is often used to store DNA and RNA while protecting it from degradation. Some protocols use TE 10:0.1 with 0.1 mM EDTA to reduce the interaction of the EDTA with downstream applications.

Solubility

Not miscible or difficult to mix in water.

Notes

200mM Tris-HCl with 20mM EDTA

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.59 g/mol
MDL Number MFCD00236359
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

pH 8.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material TE buffer

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.