Learn More
TE buffer, pH 8.0, RNAse free
Description
Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8.0 may also be used as a washing buffer. TE buffer is often used to store DNA and RNA while protecting it from degradation. Some protocols use TE 10:0.1 with 0.1 mM EDTA to reduce the interaction of the EDTA with downstream applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8.0 may also be used as a washing buffer. TE buffer is often used to store DNA and RNA while protecting it from degradation. Some protocols use TE 10:0.1 with 0.1 mM EDTA to reduce the interaction of the EDTA with downstream applications.
Solubility
Not miscible or difficult to mix in water.
Notes
200mM Tris-HCl with 20mM EDTA
Specifications
Specifications
| pH | 8.0 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Solubility Information | Not miscible or difficult to mix in water. |
| Chemical Name or Material | TE buffer, RNAse free |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.