Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tea Extract from Camellia sinensis, ≥80% theaflavins (theaflavin and theaflavin gallates) basis, MilliporeSigma™ Supelco™

Catalog No. 111004343 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mg
10 mg
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Type Analytical Standard
Quantity 10 mg
Percent Purity ≥80% theaflavins (theaflavin and theaflavin gallates) basis
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.