Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEAD1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572683
Description
TEAD1B Polyclonal Antibody for Western Blot
TEAD1 binds specifically and cooperatively to the SPH and GT-IIC enhansons (5'-GTGGAATGT-3') and activates transcription in vivo in a cell-specific manner. The activation function appears to be mediated by a limiting cell-specific transcriptional intermediary factor (TIF). It is involved in cardiac development and binds to the M-CAT motif.
Specifications
TEAD1B | |
Polyclonal | |
Unconjugated | |
tead1b | |
TEA domain family member 1 (SV40 transcriptional enhancer factor); TEA domain family member 1b; tead1; tead1b; transcriptional enhancer factor TEF-1 | |
Rabbit | |
Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
RUO | |
100006454 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
tead1b | |
KLH conjugated synthetic peptide between 324-352 amino acids from the C-terminal region of Zebrafish tead1b. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction