Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEAD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540316 Supplier Invitrogen™ Supplier No. PA540316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SGGFYGVSSQ YESLEHMTLT CSSKVCSFGK QVVEKVETER AQLEDGRFVY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 100%.

TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5'-GTGGAATGT-3'). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEAD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TEAD2
Gene Accession No. Q15562
Gene Alias Embryonic TEA domain containing factor; embryonic TEA domain-containing factor; Etdf; ETEF-1; Etf; TEA domain family member 2; TEA domain transcription factor 2; Tead2; TEAD-2; TEF4; TEF-4; transcriptional enhancer factor TEF-4
Gene Symbols TEAD2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TEAD2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8463
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.